Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002020.4 | NP_002011.2 | 27, 28 | 3773 | ||
NM_182925.4 | NP_891555.2 | 27, 28 | 3773 | |||
XM_011534478.2 | XP_011532780.1 | 27, 28 | 4151 | |||
XM_011534484.2 | XP_011532786.1 | 25, 26 | 3331 | |||
XM_017009263.1 | XP_016864752.1 | 27, 28 | 4157 | |||
XM_017009264.1 | XP_016864753.1 | 27, 28 | 4157 | |||
XM_017009265.1 | XP_016864754.1 | 27, 28 | 4158 | |||
XM_017009266.1 | XP_016864755.1 | 27, 28 | 4152 | |||
XM_017009267.1 | XP_016864756.1 | 27, 28 | 4152 | |||
XM_017009268.1 | XP_016864757.1 | 27, 28 | 3716 | |||
GenBank mRNA | AB209637.1 | - | 19, 20 | 4064 | ||
AK291679.1 | - | 27, 28 | 3749 | |||
AK309910.1 | - | 27, 28 | 3749 | |||
AY233382.1 | - | 27, 28 | 3714 | |||
AY233383.1 | - | 27, 28 | 3714 | |||
U43143.1 | - | 27, 28 | 3724 | |||
X68203.1 | - | 27, 28 | 3713 | |||
X69878.1 | - | 27, 28 | 3715 |