Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005542.4 | NP_005533.2 | 6 | 1379 | ||
NM_198336.2 | NP_938150.2 | 6 | 994 | |||
NM_198337.2 | NP_938151.1 | 4 | 1087 | |||
XM_005249542.1 | XP_005249599.1 | 7 | 1450 | |||
XM_005249543.1 | XP_005249600.1 | 7 | 1465 | |||
XM_017012177.1 | XP_016867666.1 | 6 | 1394 | |||
GenBank mRNA | AK095977.1 | - | 4 | 657 | ||
BC001880.1 | - | 6 | 1317 | 3546928 |