Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000089.3 | NP_000080.2 | 52 | 4728 | ||
GenBank mRNA | AK300194.1 | - | 11 | 950 | ||
AL833478.1 | - | 8 | 5851 | |||
BC042586.1 | - | 52 | 4375 | 4803351 | ||
BC054498.1 | - | 52 | 4357 | 6474549 | ||
J03464.1 | - | 52 | 4726 | |||
Z74616.1 | - | 52 | 4396 |