Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000090.3 | NP_000081.1 | 10, 11 | 924 | ||
GenBank mRNA | AK308010.1 | - | 6, 7 | 689 | ||
BC028178.1 | - | 10, 11 | 922 | 5405119 | ||
X14420.1 | - | 10, 11 | 909 | |||
X15332.1 | - | 6, 7 | 366 |