Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000090.3 | NP_000081.1 | 51 | 4528 | ||
GenBank mRNA | AK091853.1 | - | 3 | 1790 | ||
BC028178.1 | - | 41 | 3617 | 5405119 | ||
X01742.1 | - | 4 | 823 | |||
X06700.1 | - | 12 | 1564 | |||
X14420.1 | - | 51 | 4513 |