Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000091.4 | NP_000082.2 | 52 | 6709 | ||
XM_005246277.3 | XP_005246334.1 | 51 | 6580 | |||
XM_011510556.2 | XP_011508858.1 | 39 | 5741 |