Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000092.4 | NP_000083.3 | 18 | 1725 | ||
XM_005246281.3 | XP_005246338.1 | 18 | 1259 | |||
XM_006712246.3 | XP_006712309.1 | 18 | 1397 | |||
XM_011510557.2 | XP_011508859.1 | 18 | 1397 | |||
XM_011510558.2 | XP_011508860.1 | 18 | 1397 | |||
XM_011510559.2 | XP_011508861.1 | 18 | 1397 | |||
XM_011510560.2 | XP_011508862.1 | 18 | 1397 | |||
XM_011510561.2 | XP_011508863.1 | 18 | 1397 | |||
XM_011510562.2 | XP_011508864.1 | 18 | 1397 | |||
XM_011510565.2 | XP_011508867.1 | 18 | 1397 | |||
XM_011510566.2 | XP_011508868.1 | 18 | 1397 | |||
XM_011510567.2 | XP_011508869.1 | 18 | 1397 | |||
XM_011510568.2 | XP_011508870.1 | 18 | 1397 | |||
XM_011510569.2 | XP_011508871.1 | 18 | 1397 | |||
XM_011510570.2 | XP_011508872.1 | 18 | 1397 | |||
XM_017003296.1 | XP_016858785.1 | 18 | 1397 | |||
XM_017003297.1 | XP_016858786.1 | 18 | 1397 | |||
XM_017003298.1 | XP_016858787.1 | 18 | 1397 | |||
XM_017003299.1 | XP_016858788.1 | 18 | 1397 | |||
XM_017003300.1 | XP_016858789.1 | 18 | 1397 | |||
GenBank mRNA | X81053.1 | - | 18 | 1279 |