Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000092.4 | NP_000083.3 | 46 | 5166 | ||
XM_005246281.3 | XP_005246338.1 | 46 | 4700 | |||
XM_005246282.3 | XP_005246339.1 | 40 | 4283 | |||
XM_011510557.2 | XP_011508859.1 | 47 | 4757 | |||
XM_011510558.2 | XP_011508860.1 | 45 | 4730 | |||
XM_011510559.2 | XP_011508861.1 | 46 | 4838 | |||
XM_011510560.2 | XP_011508862.1 | 46 | 4838 | |||
XM_011510561.2 | XP_011508863.1 | 46 | 4838 | |||
XM_011510562.2 | XP_011508864.1 | 46 | 4838 | |||
XM_011510572.2 | XP_011508874.1 | 25 | 3063 | |||
XM_017003296.1 | XP_016858785.1 | 46 | 4838 | |||
XM_017003297.1 | XP_016858786.1 | 46 | 4721 | |||
XM_017003298.1 | XP_016858787.1 | 46 | 4838 | |||
XM_017003299.1 | XP_016858788.1 | 46 | 4838 | |||
GenBank mRNA | D17391.1 | - | 8 | 859 | ||
X81053.1 | - | 46 | 4720 |