Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL7A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000094.3 | NP_000085.1 | 22 | 2885 | ||
XM_011533337.1 | XP_011531639.1 | 23 | 2920 | |||
XM_017005688.1 | XP_016861177.1 | 23 | 2920 | |||
XM_017005689.1 | XP_016861178.1 | 23 | 2920 | |||
XM_017005690.1 | XP_016861179.1 | 23 | 2920 | |||
XM_017005691.1 | XP_016861180.1 | 23 | 2920 | |||
XM_017005692.1 | XP_016861181.1 | 23 | 2920 | |||
GenBank mRNA | D13694.1 | - | 18 | 2299 | ||
L02870.1 | - | 22 | 2997 | |||
M65158.1 | - | 4 | 442 | |||
M96984.1 | - | 14 | 1780 | |||
S51236.1 | - | 14 | 1780 |