Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIVM-ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204425.1 | NP_001191354.1 | 16 | 2660 | ||
GenBank mRNA | AB209312.1 | - | 16 | 2452 | ||
LM994822.1 | - | 16 | 2538 |
Gene Symbol: ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000123.3 | NP_000114.2 | 8 | 1602 | ||
GenBank mRNA | AF462447.1 | - | 8 | 1559 | ||
AK294708.1 | - | 8 | 1531 | |||
AK299758.1 | - | 8 | 1314 | |||
AK303558.1 | - | 9 | 1505 | |||
BC031522.2 | - | 8 | 1588 | 3869893 | ||
BX647399.1 | - | 8 | 1431 | |||
D16305.1 | - | 8 | 1186 | |||
L20046.1 | - | 8 | 1224 | |||
LM994821.1 | - | 4 | 672 | |||
X69978.1 | - | 8 | 1373 |