Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000393.3 | NP_000384.2 | 52, 53 | 4388 | ||
XM_011510573.2 | XP_011508875.1 | 52, 53 | 4170 | |||
GenBank mRNA | AB209045.1 | - | 52, 53 | 4250 | ||
AK300146.1 | - | 4, 5 | 865 | |||
BC043613.1 | - | 14, 15 | 1483 | |||
BC086874.1 | - | 36, 37 | 3377 | 30708812 | ||
M11718.1 | - | 2, 3 | 426 | |||
X04758.1 | - | 34, 35 | 2913 | |||
Y14690.1 | - | 52, 53 | 4242 |