Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000393.3 | NP_000384.2 | 54 | 5656 | ||
XM_011510573.2 | XP_011508875.1 | 54 | 5438 | |||
GenBank mRNA | AB209045.1 | - | 54 | 5518 |