Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000396.3 | NP_000387.1 | 8 | 1324 | ||
GenBank mRNA | AK130134.1 | - | 8 | 1143 | ||
AK308229.1 | - | 7 | 985 | |||
BC016058.1 | - | 8 | 1206 | 4860926 | ||
S79895.1 | - | 8 | 1236 | |||
U13665.1 | - | 8 | 1200 | |||
U20280.1 | - | 8 | 1113 |