Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004286.4 | NP_004277.2 | 12 | 3120 | ||
GenBank mRNA | AF370385.1 | - | 1 | 693 | ||
AK095950.1 | - | 1 | 681 | |||
AK096647.1 | - | 2 | 2458 | |||
BC014075.1 | - | 12 | 2960 |