Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004288.4 | NP_004279.3 | 5 | 501 | ||
XM_017005386.1 | XP_016860875.1 | 5 | 692 | |||
GenBank mRNA | AF068836.1 | - | 5 | 462 | ||
AK301544.1 | - | 8 | 594 | |||
AK312753.1 | - | 5 | 498 | |||
BC036449.2 | - | 5 | 459 | 4830272 | ||
CD642163.1 | - | 5 | 554 | |||
DB219691.1 | - | 5 | 502 | |||
L06633.1 | - | 5 | 415 |