Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000560.3 | NP_000551.1 | 8 | 903 | ||
NM_001040033.1 | NP_001035122.1 | 9 | 908 | |||
NM_001320638.1 | NP_001307567.1 | 6 | 726 | |||
GenBank mRNA | AK298723.1 | - | 6 | 673 | ||
BC035456.1 | - | 9 | 890 | |||
BC040693.1 | - | 8 | 833 | 5498854 | ||
M37033.1 | - | 8 | 830 | |||
M60871.1 | - | 8 | 810 |