Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000567.2 | NP_000558.2 | 2 | 752 | ||
XM_011509207.2 | XP_011507509.1 | 2 | 786 | |||
GenBank mRNA | AK289443.1 | - | 2 | 752 | ||
AK313624.1 | - | 2 | 752 | |||
BC020766.1 | - | 3 | 353 | |||
BC070257.1 | - | 2 | 513 | |||
BC125135.1 | - | 2 | 680 | 40119392 | ||
BG618397.1 | - | 1 | 216 | |||
X56214.1 | - | 2 | 737 | |||
X56692.1 | - | 2 | 737 |