Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000726.3 | NP_000717.2 | 10 | 879 | ||
NM_001005746.2 | NP_001005746.1 | 10 | 829 | |||
NM_001005747.2 | NP_001005747.1 | 9 | 967 | |||
NM_001145798.1 | NP_001139270.1 | 10 | 879 | |||
NM_001320722.1 | NP_001307651.1 | 10 | 1127 | |||
XM_011511795.2 | XP_011510097.1 | 10 | 2596 | |||
XM_011511796.2 | XP_011510098.1 | 9 | 1550 | |||
XM_017004883.1 | XP_016860372.1 | 11 | 2418 | |||
XM_017004884.1 | XP_016860373.1 | 9 | 2252 | |||
XM_017004885.1 | XP_016860374.1 | 8 | 2092 | |||
GenBank mRNA | AB302276.1 | - | 10 | 823 | ||
AF038852.1 | - | 10 | 789 | |||
AK290049.1 | - | 9 | 948 | |||
AK291378.1 | - | 11 | 962 | |||
AK294398.1 | - | 10 | 803 | |||
AK316045.1 | - | 10 | 820 | |||
BC075049.2 | - | 10 | 830 | 30915578 | ||
U95020.1 | - | 10 | 879 |