Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCKAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000730.2 | NP_000721.1 | 2 | 352 | ||
GenBank mRNA | AK313978.1 | - | 2 | 394 | ||
AY322549.1 | - | 2 | 199 | |||
BC074987.2 | - | 2 | 247 | 30915519 | ||
L13605.1 | - | 2 | 270 | |||
L19315.1 | - | 2 | 352 |
Set Membership: |
G-protein coupled receptor |