Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000767.4 | NP_000758.1 | 1, 2 | 185 | ||
XM_005258569.4 | XP_005258626.1 | 1, 2 | 281 | |||
XM_006723050.3 | XP_006723113.1 | 1, 2 | 281 | |||
XM_011526546.2 | XP_011524848.1 | 1, 2 | 284 | |||
XM_011526547.2 | XP_011524849.1 | 1, 2 | 282 | |||
XM_011526548.2 | XP_011524850.1 | 1, 2 | 278 | |||
GenBank mRNA | AF182277.1 | - | 1, 2 | 180 | ||
BC067430.1 | - | 1, 2 | 185 | 7002137 | ||
BC067431.1 | - | 1, 2 | 185 | 7002142 | ||
M29874.1 | - | 1, 2 | 184 | |||
X13494.1 | - | 1, 2 | 117 |
Gene Symbol: CYP2B7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001278.1 | 1, 2 | 186 | |||
GenBank mRNA | AK307933.1 | - | 1, 2 | 232 | ||
AK310944.1 | - | 1, 2 | 236 | |||
DQ198366.1 | - | 1, 2 | 178 | |||
DQ198367.1 | - | 1, 2 | 178 |