Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000767.4 | NP_000758.1 | 9 | 1662 | ||
XM_005258569.4 | XP_005258626.1 | 8 | 1616 | |||
XM_011526548.2 | XP_011524850.1 | 6 | 1275 | |||
XM_011526549.2 | XP_011524851.1 | 7 | 1217 | |||
XM_011526550.2 | XP_011524852.1 | 5 | 1070 | |||
GenBank mRNA | AK301620.1 | - | 5 | 1059 | ||
M29874.1 | - | 9 | 1661 | |||
X06399.1 | - | 3 | 1007 | |||
X06400.1 | - | 3 | 606 |