Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000778.3 | NP_000769.2 | 12 | 2147 | ||
NM_001319155.1 | NP_001306084.1 | 12 | 2051 | |||
NR_134988.1 | 10 | 1801 | ||||
NR_134989.1 | 11 | 1992 | ||||
NR_134990.1 | 11 | 1986 | ||||
NR_134991.1 | 11 | 1973 | ||||
NR_134992.1 | 8 | 1602 | ||||
NR_134993.1 | 9 | 1736 | ||||
NR_134994.1 | 11 | 2008 | ||||
XM_017000464.1 | XP_016855953.1 | 10 | 1845 | |||
XM_017000465.1 | XP_016855954.1 | 12 | 2066 | |||
GenBank mRNA | D26481.1 | - | 12 | 2110 | ||
L04751.1 | - | 12 | 2128 | |||
S67580.1 | - | 12 | 2138 | |||
S67581.1 | - | 12 | 2137 |
Gene Symbol: CYP4A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005270767.3 | XP_005270824.1 | 10 | 1845 |