Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003954.2 | NP_001003954.1 | 12 | 1523 | ||
NM_004306.3 | NP_004297.2 | 11 | 1400 | |||
GenBank mRNA | AJ306450.1 | - | 12 | 1499 | ||
AK223374.1 | - | 11 | 1399 | |||
AK290281.1 | - | 12 | 1509 | |||
Z11502.1 | - | 11 | 1387 |