Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001234.4 | NP_001225.1 | 3 | 1262 | ||
NM_033337.2 | NP_203123.1 | 2 | 1358 | |||
GenBank mRNA | AF043101.1 | - | 2 | 1273 | ||
Y14747.1 | - | 3 | 1200 |