Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220488.1 | NP_001207417.1 | 7 | 1136 | ||
NM_001220489.1 | NP_001207418.1 | 5 | 913 | |||
NM_001220490.1 | NP_001207419.1 | 5 | 821 | |||
NM_001257.4 | NP_001248.1 | 6 | 1030 | |||
XM_011522804.2 | XP_011521106.1 | 5 | 637 | |||
XM_017022848.1 | XP_016878337.1 | 7 | 1053 | |||
GenBank mRNA | AK295886.1 | - | 5 | 651 | ||
AK298612.1 | - | 5 | 739 | |||
AK304733.1 | - | 7 | 891 | |||
AK307862.1 | - | 7 | 827 | |||
BC028624.2 | - | 6 | 822 | 4825734 | ||
BC030653.2 | - | 6 | 822 | 4811561 | ||
BU162665.1 | - | 2 | 163 | |||
CN263227.1 | - | 1 | 54 | |||
DB003683.1 | - | 1 | 101 | |||
L34058.1 | - | 6 | 1175 | |||
U59288.1 | - | 6 | 947 |