Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220488.1 | NP_001207417.1 | 12 | 2059 | ||
NM_001220489.1 | NP_001207418.1 | 10 | 1836 | |||
NM_001220490.1 | NP_001207419.1 | 10 | 1744 | |||
NM_001257.4 | NP_001248.1 | 11 | 1953 | |||
XM_011522804.2 | XP_011521106.1 | 10 | 1560 | |||
GenBank mRNA | AK294277.1 | - | 5 | 816 | ||
AK295886.1 | - | 10 | 1574 | |||
AK298612.1 | - | 10 | 1662 | |||
AK304592.1 | - | 4 | 452 | |||
AK304733.1 | - | 12 | 1814 | |||
BC028624.2 | - | 11 | 1745 | 4825734 | ||
BC030653.2 | - | 11 | 1745 | 4811561 | ||
L34058.1 | - | 11 | 2098 | |||
U59288.1 | - | 11 | 1870 |