Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220488.1 | NP_001207417.1 | 15 | 3544 | ||
NM_001220489.1 | NP_001207418.1 | 13 | 3321 | |||
NM_001220490.1 | NP_001207419.1 | 13 | 3229 | |||
NM_001257.4 | NP_001248.1 | 14 | 3438 | |||
XM_011522804.2 | XP_011521106.1 | 13 | 3045 | |||
GenBank mRNA | BC028624.2 | - | 14 | 3230 | 4825734 | |
BC030653.2 | - | 14 | 3230 | 4811561 | ||
CB321814.1 | - | 1 | 408 | |||
U59288.1 | - | 14 | 3352 |