Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265.4 | NP_001256.3 | 3 | 1827 | ||
XM_011534875.2 | XP_011533177.1 | 3 | 1820 | |||
XM_011534878.2 | XP_011533180.1 | 3 | 1982 | |||
XM_011534879.2 | XP_011533181.1 | 3 | 1947 | |||
GenBank mRNA | BC014461.1 | - | 3 | 1639 | 4867859 | |
U51096.1 | - | 3 | 1521 |