Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278.4 | NP_001269.3 | 20, 21 | 2279 | ||
NM_001320928.1 | NP_001307857.1 | 20, 21 | 2262 | |||
GenBank mRNA | AB209028.1 | - | 19, 20 | 4562 | ||
AF009225.1 | - | 20, 21 | 2236 | |||
AF012890.1 | - | 20, 21 | 2201 | |||
AF080157.1 | - | 20, 21 | 2296 | |||
AL133012.1 | - | 2, 3 | 3578 | |||
BC092514.1 | - | 20, 21 | 2217 | 3063613 | ||
U22512.1 | - | 19, 20 | 2109 |