Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIRBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280.2 | NP_001271.1 | 7 | 1074 | ||
NM_001300829.1 | NP_001287758.1 | 6 | 1451 | |||
NR_023312.2 | 6 | 1510 | ||||
NR_023313.2 | 5 | 1342 | ||||
XM_006722637.1 | XP_006722700.1 | 7 | 1137 | |||
GenBank mRNA | AF021336.1 | - | 7 | 1006 | ||
AK094781.1 | - | 5 | 1275 | |||
AK095781.1 | - | 4 | 1569 | |||
AK128423.1 | - | 5 | 1363 | |||
AK304470.1 | - | 6 | 1283 | |||
BC000403.2 | - | 7 | 986 | 2822968 | ||
BC000901.1 | - | 7 | 991 | 2900585 | ||
D78134.1 | - | 7 | 1004 |