Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281.2 | NP_001272.2 | 3 | 906 | ||
NM_001300971.1 | NP_001287900.1 | 3 | 318 | |||
GenBank mRNA | AF013488.1 | - | 3 | 396 | ||
BC005969.1 | - | 3 | 383 | 4076810 | ||
BC052812.1 | - | 3 | 406 | 6527446 | ||
BQ923739.1 | - | 3 | 319 | |||
BT007424.1 | - | 3 | 331 | |||
CR541971.1 | - | 2 | 178 | |||
CR996176.1 | - | 3 | 318 | |||
D49738.1 | - | 3 | 451 |