Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004315.5 | NP_004306.3 | 4 | 475 | ||
NM_177924.4 | NP_808592.2 | 4 | 542 | |||
XM_005273504.3 | XP_005273561.1 | 4 | 303 | |||
GenBank mRNA | AK025732.1 | - | 5 | 445 | ||
AK098299.1 | - | 5 | 265 | |||
AK222780.1 | - | 5 | 278 | |||
AK289481.1 | - | 5 | 443 | |||
AY305384.1 | - | 4 | 316 | |||
BC016481.1 | - | 5 | 265 | 3923451 | ||
BG714023.1 | - | 4 | 288 | |||
DA260852.1 | - | 4 | 475 | |||
DA306573.1 | - | 5 | 542 | |||
U47674.1 | - | 4 | 246 | |||
U70063.1 | - | 5 | 247 |