Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311.4 | NP_001302.1 | 5 | 340 | ||
GenBank mRNA | AK092741.1 | - | 2 | 2104 | ||
AK098290.1 | - | 1 | 1307 | |||
BC002738.2 | - | 4 | 274 | 3631097 | ||
BQ128075.1 | - | 5 | 345 | |||
BU500642.1 | - | 4 | 284 | |||
U09770.1 | - | 4 | 290 | |||
U58630.1 | - | 4 | 226 |