Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSE1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256135.1 | NP_001243064.1 | 24 | 3102 | ||
NM_001316.3 | NP_001307.2 | 25 | 3270 | |||
NR_045796.1 | 22 | 2785 | ||||
XM_011528599.2 | XP_011526901.1 | 25 | 3377 | |||
GenBank mRNA | AF053641.1 | - | 25 | 3204 | ||
AF053642.1 | - | 6 | 1053 | |||
BC109313.2 | - | 25 | 3183 | 40036129 | ||
BC109314.2 | - | 25 | 3183 | 40036130 |