Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CX3CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171171.1 | NP_001164642.1 | 2 | 610 | ||
NM_001171172.1 | NP_001164643.1 | 2 | 570 | |||
NM_001171174.1 | NP_001164645.1 | 2 | 506 | |||
NM_001337.3 | NP_001328.1 | 2 | 462 | |||
GenBank mRNA | AK225764.1 | - | 2 | 463 | ||
AK307911.1 | - | 3 | 262 | |||
AK312373.1 | - | 2 | 461 | |||
BC028078.1 | - | 2 | 446 | 5216452 | ||
DA396634.1 | - | 2 | 452 | |||
U20350.1 | - | 2 | 457 | |||
U28934.1 | - | 2 | 415 |