Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080744.1 | NP_001074213.1 | 23, 24 | 2735 | ||
NM_001080745.1 | NP_001074214.1 | 23, 24 | 2693 | |||
NM_001346.2 | NP_001337.2 | 24, 25 | 2810 | |||
GenBank mRNA | AK304440.1 | - | 22, 23 | 2631 | ||
AK314192.1 | - | 24, 25 | 2755 | |||
AY737485.1 | - | 22, 23 | 2388 | |||
BC089411.1 | - | 23, 24 | 2489 | 3081161 | ||
BC112363.1 | - | 22, 23 | 2416 | 40054027 | ||
D26135.1 | - | 24, 25 | 2789 |