Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAXX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141969.1 | NP_001135441.1 | 6 | 1778 | ||
NM_001141970.1 | NP_001135442.1 | 6 | 1709 | |||
NM_001254717.1 | NP_001241646.1 | 5 | 1519 | |||
NM_001350.4 | NP_001341.1 | 6 | 1775 | |||
XM_005248860.3 | XP_005248917.1 | 4 | 625 | |||
GenBank mRNA | AB209493.1 | - | 2 | 2243 | ||
AF006041.1 | - | 1 | 96 | |||
AF015956.2 | - | 5 | 1625 | |||
AF039136.1 | - | 6 | 1718 | |||
AF050179.1 | - | 6 | 1686 | |||
AF097742.1 | - | 5 | 1578 | |||
AK223404.1 | - | 6 | 1776 | |||
AK292187.1 | - | 6 | 1776 | |||
AK303767.1 | - | 6 | 1652 | |||
AK303854.1 | - | 5 | 1476 | |||
AK313034.1 | - | 6 | 1718 | |||
BC000220.1 | - | 573 | ||||
BC109073.1 | - | 6 | 1661 | 40034214 | ||
BC109074.1 | - | 6 | 1661 | 40034219 | ||
CR457085.1 | - | 5 | 1571 | |||
HQ436528.1 | - | 5 | 1571 | |||
HQ436529.1 | - | 5 | 1571 |