Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAXX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141969.1 | NP_001135441.1 | 8 | 2516 | ||
NM_001141970.1 | NP_001135442.1 | 8 | 2447 | |||
NM_001254717.1 | NP_001241646.1 | 7 | 2257 | |||
NM_001350.4 | NP_001341.1 | 8 | 2513 | |||
XM_005248860.3 | XP_005248917.1 | 6 | 1363 | |||
GenBank mRNA | AB209493.1 | - | 2 | 3335 | ||
AF006041.1 | - | 3 | 834 | |||
AF015956.2 | - | 7 | 2363 | |||
AF039136.1 | - | 8 | 2456 | |||
AF050179.1 | - | 8 | 2424 | |||
AF097742.1 | - | 7 | 2316 | |||
AK223404.1 | - | 8 | 2514 | |||
AK292187.1 | - | 8 | 2514 | |||
AK303767.1 | - | 8 | 2390 | |||
BC000220.1 | - | 1514 |