Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126111.2 | NP_001119583.1 | 6 | 2621 | ||
NM_004337.2 | NP_004328.1 | 6 | 2575 | |||
XM_011517287.2 | XP_011515589.1 | 6 | 2483 | |||
XM_011517288.2 | XP_011515590.1 | 3 | 2308 | |||
GenBank mRNA | AF061326.1 | - | 6 | 2574 |