Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128827.2 | NP_001122299.1 | 10 | 1570 | ||
NM_001321074.1 | NP_001308003.1 | 12 | 2418 | |||
NM_001321075.1 | NP_001308004.1 | 10 | 1579 | |||
NM_001321076.1 | NP_001308005.1 | 9 | 1224 | |||
NM_001321077.1 | NP_001308006.1 | 9 | 1215 | |||
NM_001365.4 | NP_001356.1 | 12 | 2427 | |||
NR_135527.1 | 12 | 2427 | ||||
XM_005256491.1 | XP_005256548.1 | 10 | 1669 | |||
XM_011523698.1 | XP_011522000.1 | 12 | 2315 | |||
XM_011523699.2 | XP_011522001.1 | 13 | 1557 | |||
XM_011523702.1 | XP_011522004.1 | 9 | 1094 | |||
XM_017024288.1 | XP_016879777.1 | 9 | 1315 | |||
XM_017024289.1 | XP_016879778.1 | 9 | 1306 | |||
XM_017024290.1 | XP_016879779.1 | 9 | 1784 | |||
GenBank mRNA | AK293835.1 | - | 10 | 1576 | ||
AK296357.1 | - | 12 | 2115 | |||
AK297346.1 | - | 9 | 1172 | |||
AK299801.1 | - | 9 | 1186 | |||
AK308533.1 | - | 9 | 1329 | |||
AK316096.1 | - | 10 | 1132 | |||
BC040533.1 | - | 4 | 572 | |||
BC136553.1 | - | 12 | 1951 | 9020543 | ||
U83192.1 | - | 12 | 2085 |