Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTTG1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286822.1 | NP_001273751.1 | 3 | 2429 | ||
NM_004339.3 | NP_004330.1 | 6 | 2757 | |||
NR_104597.1 | 5 | 2648 | ||||
GenBank mRNA | AF149785.1 | - | 6 | 2702 | ||
AK094882.1 | - | 7 | 2462 | |||
AK095586.1 | - | 1 | 3058 | |||
BC000415.2 | - | 6 | 2515 | 2961420 | ||
BC012858.1 | - | 6 | 2541 | 3857505 | ||
BC019295.1 | - | 6 | 2504 | 2823814 | ||
BC020983.1 | - | 6 | 2520 | 3851447 | ||
BC031097.1 | - | 7 | 2484 | 5288388 | ||
BC034250.1 | - | 6 | 2529 | 3862830 | ||
L48984.1 | - | 3 | 2154 | |||
Z50022.1 | - | 6 | 2560 |