Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001406.3 | NP_001397.1 | 5 | 1422 | ||
GenBank mRNA | AK027329.1 | - | 2 | 545 | ||
BC022499.1 | - | 4 | 861 | |||
BC042944.2 | - | 5 | 1422 | 5286243 | ||
U57001.1 | - | 5 | 1422 | |||
U62775.1 | - | 5 | 1226 | |||
U66406.1 | - | 5 | 1638 |