Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001408.2 | NP_001399.1 | 1 | 1881 | ||
XM_005270580.4 | XP_005270637.1 | 1 | 2195 | |||
GenBank mRNA | AF234887.1 | - | 1 | 1882 | ||
D87469.2 | - | 1 | 1613 |
Set Membership: |
G-protein coupled receptor |