Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001415.3 | NP_001406.1 | 2 | 139 | ||
GenBank mRNA | AB451230.1 | - | 2 | 118 | ||
AB451353.1 | - | 2 | 118 | |||
AK222577.1 | - | 2 | 131 | |||
AK312247.1 | - | 2 | 131 | |||
BC019906.1 | - | 2 | 139 | 4419438 | ||
L19161.1 | - | 2 | 118 |
Gene Symbol: LOC255308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC087847.1 | - | 1 | 116 |