Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127197.1 | NP_001120669.1 | 6 | 863 | ||
NM_001421.3 | NP_001412.1 | 6 | 945 | |||
XM_005262389.3 | XP_005262446.1 | 6 | 1281 | |||
XM_011531307.2 | XP_011529609.1 | 5 | 691 | |||
XM_011531308.2 | XP_011529610.1 | 4 | 473 | |||
GenBank mRNA | AF000670.1 | - | 5 | 662 | ||
AK314972.1 | - | 6 | 862 | |||
BC017194.2 | - | 6 | 828 | 3138355 | ||
U32645.1 | - | 6 | 942 |