Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001430.4 | NP_001421.2 | 4 | 893 | ||
XM_011532698.2 | XP_011531000.1 | 4 | 651 | |||
GenBank mRNA | BC051338.1 | - | 4 | 698 | 6305604 | |
U51626.1 | - | 4 | 415 | |||
U81984.1 | - | 4 | 532 |