Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001432.2 | NP_001423.1 | 5 | 3703 | ||
GenBank mRNA | D30783.1 | - | 5 | 3702 |