Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001436.3 | NP_001427.2 | 7, 8 | 917 | ||
XM_005258651.2 | XP_005258708.1 | 7, 8 | 895 | |||
XM_011526623.2 | XP_011524925.1 | 6, 7 | 698 | |||
GenBank mRNA | AB451384.1 | - | 6, 7 | 803 | ||
AK296950.1 | - | 7, 8 | 882 | |||
BC015218.2 | - | 5, 6 | 622 | |||
BC019260.1 | - | 7, 8 | 871 | 3504198 | ||
BC019609.1 | - | 5, 6 | 621 | |||
BT006830.1 | - | 6, 7 | 803 | |||
BT020144.1 | - | 6, 7 | 803 | |||
CR457069.1 | - | 6, 7 | 803 | |||
M59849.1 | - | 7, 8 | 831 | |||
X56597.1 | - | 7, 8 | 862 |