Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040275.1 | NP_001035365.1 | 2 | 557 | ||
NM_001214902.1 | NP_001201831.1 | 1 | 179 | |||
NM_001271876.1 | NP_001258805.1 | 1 | 179 | |||
NM_001271877.1 | NP_001258806.1 | 1 | 179 | |||
NM_001291712.1 | NP_001278641.1 | 7 | 1817 | |||
NM_001291723.1 | NP_001278652.1 | 2 | 832 | |||
NM_001437.2 | NP_001428.1 | 2 | 557 | |||
NR_073496.1 | 2 | 832 | ||||
NR_073497.1 | 1 | 179 | ||||
XM_017021079.1 | XP_016876568.1 | 3 | 1221 | |||
XM_017021080.1 | XP_016876569.1 | 3 | 2030 | |||
XM_017021081.1 | XP_016876570.1 | 3 | 1126 | |||
XM_017021082.1 | XP_016876571.1 | 3 | 978 | |||
XM_017021083.1 | XP_016876572.1 | 2 | 395 | |||
XM_017021084.1 | XP_016876573.1 | 3 | 1221 | |||
GenBank mRNA | AB006589.1 | - | 7 | 1364 | ||
AB006590.1 | - | 1 | 187 | |||
AB209620.1 | - | 1 | 185 | |||
AF051427.1 | - | 2 | 507 | |||
AF051428.1 | - | 2 | 507 | |||
AF060555.1 | - | 2 | 559 | |||
AF124790.1 | - | 1 | 140 | |||
AK292370.1 | - | 2 | 394 | |||
BC024181.2 | - | 2 | 374 | 4825937 | ||
DQ777076.1 | - | 1 | 89 | |||
DQ777077.1 | - | 1 | 89 | |||
DQ838582.1 | - | 1 | 89 | |||
DQ838583.1 | - | 1 | 89 | |||
HQ692820.1 | - | 1 | 89 | |||
HQ692821.1 | - | 1 | 89 | |||
KJ534833.1 | - | 1 | 89 |