Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001447.2 | NP_001438.1 | 1 | 472 | ||
XM_006714761.3 | XP_006714824.1 | 2 | 1043 | |||
XM_011537600.2 | XP_011535902.1 | 4 | 811 | |||
XM_011537603.2 | XP_011535905.1 | 2 | 744 | |||
XM_017009224.1 | XP_016864713.1 | 4 | 813 | |||
XM_017009225.1 | XP_016864714.1 | 3 | 655 | |||
XM_017009226.1 | XP_016864715.1 | 2 | 1048 | |||
XM_017009227.1 | XP_016864716.1 | 2 | 1047 | |||
GenBank mRNA | AF231022.1 | - | 1 | 472 | ||
AK308619.1 | - | 2 | 637 |